Size exclusion chromatography columns, Bio SEC-3 and Bio SEC-5
: Agilent
Bio SEC-3 columns are packed with spherical, narrowly dispersed 3 µm silica particles coated with a proprietary hydrophilic layer for high recovery and minimal secondary interactions which provides more consistent separations.
- Compatible with most aqueous buffers
- Stable under high and low salt condition
Bio SEC- 5 columns are packed with 5 µm silica particles coated with a proprietary, neutral, hydrophilic layer for maximum efficiency and stability.
- Catalog No:
- HEWL5190-2544
- HEWL5190-6848
- HEWL5190-2532
- HEWL5190-2511
- HEWL5190-2510
- HEWL5190-2521
- HEWL5190-2533
- HEWL5190-2513
- HEWL5190-2512
- HEWL5190-2534
- HEWL5190-2538
- HEWL5190-2526
- HEWL5190-2525
- HEWL5190-2537
- HEWL5190-2524
- HEWL5190-2536
- HEWL5190-2535
- HEWL5190-2530
- HEWL5190-2542
- HEWL5190-2529
- HEWL5190-2541
- HEWL5190-2528
- HEWL5190-2540
- HEWL5190-2527
- HEWL5190-2539
- HEWL5190-2519
- HEWL5190-2506
- HEWL5190-2505
- HEWL5190-2523
- HEWL5190-2522
- HEWL5190-2507
- HEWL5190-2545
- HEWL5190-2520
- HEWL5190-2531
- HEWL5190-6847
- HEWL5190-2543
- HEWL5190-2509
- HEWL5190-2514
- HEWL5190-2508
- HEWL5190-2518
- HEWL5190-2503
- HEWL5190-2502
- HEWL5190-2517
- HEWL5190-2501
- HEWL5190-2516
- HEWL5190-2515
- HEWL5190-6846
- HEWL5190-2504
- Max. pressure:
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- 2000 psi
- pH range:
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- 2 - 8,5
- Temperature limit:
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
- 10...20 °C
Frequently Bought Together