ContactUsLinkComponent
Untitled-6-Recovered.jpg
WebSeal Well Plates by Thermo Scientific
A comprehensive range of PP well plates.
Our plate portfolio contains well plates which meets the requirements of today’s chromatographer using autosampler for plates. Compatible with a broad range of instruments, our well plate range offers solutions from simple routine analyses to specific and challenging sample handling problems.
    Handle More Samples in Less Time
    • Low volume and high recovery cavity
    Solutions for all Applications
    • Chromatography tested for solvent compatibility and inertness
    Autosampler Compatible
    • Using ANSI/SBS standards
    Product selection
    Thermo Scientific Webseal_1.jpg
    Low cost, standard performance commodity plates, seals and tapes (GC and LC tested and confirmed to be acceptable).
    Thermo Scientific Webseal_2.jpg
    Certified Plates and Mats
    Certified products, low bleed plates & high performance mats (GC/MS and LC/MS tested and confirmed to be excellent).