Orders LinkContactUsLinkComponent
InletLiners-1-955x690.jpg
Inlet Liners
Maintaining a clean injection port is critical to an inert flow path and reliable results. This involves the use of glass inlet liners, precision inlet seals, and reliable O-rings.
Agilent offers a wide variety of inlet liners for split splitless injections, cool on-column (COC) injections and multimode injections, with and without glass wool. If your samples require complete inertness, try any of the products coated with Agilent's exclusive Ultra Inert technology. We also provide a complete offering of liner O-rings and inlet liners for third-party instruments, making Agilent a one-stop shop for all your supplies.
DimpledLinersImage-400x495.jpg
The 2 mm dimpled liner to be used in the Agilent MultiMode Inlet when analyzing heavy matrix samples such as food or environmental extracts.
DirectConnectLinersPicture-400x495.jpg
Direct Connect liners provide maximum recovery and minimal decomposition of active compounds for methods requiring splitless injection.
FlipTopSealingSystem-400x495.jpg
Flip-top inlet sealing system allows you to safely and reliably change an inlet liner in as little as 30 seconds,in a consistent leak-free manner.
InletConvenienceKits-400x495.jpg
Convenience kits are an easy way to get all the supplies you need using one part number.PM kits include septa, liners, O-rings, gold seals and traps.
LinerORing-400x495.jpg
Liner O-rings are used to form a proper seal around the outside of the inlet liner between the inlet weldment and the turn-top. O-rings are composed of either graphite or fluorocarbon material, graphite O-rings are used when operating inlet temperatures exceed 350°C.
SplitLiners-400x495.jpg
Split injections are intended for use with a split/splitless injection port in gas chromatography analysis.
SplitSplitlessInletGoldSeals-400x495.jpg
The gold seal forms a leak-free seal with the bottom of the inlet body and column ferrule to minimize both sample contamination and sample loss
SplitlessLiners-400x495.jpg
Choose a liner without glass wool if your sample contains nonvolatiles or analytes within a wide boiling range.
UltraInertLiners-400x495.jpg
Agilent Ultra Inert inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface.