Order Entry
Slovenia
ContactUsLinkComponent
20mm headspace cap and septa preassembled. 1000 caps with septa inserted 0.005 thick natural ptfe / 0.120 ultra low bleed white silicone (0.125 total thickness) septaregular crimp top cap and septa without pressure release feature septa is condit 1 * 1.000 items
Catalog # PERCN9302977
Supplier:  PerkinElmer
New Product
20mm headspace cap and septa preassembled. 1000 caps with septa inserted 0.005 thick natural ptfe / 0.120 ultra low bleed white silicone (0.125 total thickness) septaregular crimp top cap and septa without pressure release feature septa is condit 1 * 1.000 items
Catalog # PERCN9302977
Supplier:  PerkinElmer