Order Entry
Slovenia
ContactUsLinkComponent
QTL28765B FLAT PACK HYGIENOX TABLE 1200X600 ELECTROPOLISHED Manufactured from 304L stainless steel Electropolished finish for lower Ra Overall 1200 x 600 x 840mm high Supplied flat packed for room access Tubular frame & legs ESD swivel castors, 2 bra 1 * 1 items
Catalog # TEKMAA004-B
Supplier:  TEKNOMEK
New Product
QTL28765B FLAT PACK HYGIENOX TABLE 1200X600 ELECTROPOLISHED Manufactured from 304L stainless steel Electropolished finish for lower Ra Overall 1200 x 600 x 840mm high Supplied flat packed for room access Tubular frame & legs ESD swivel castors, 2 bra 1 * 1 items
Catalog # TEKMAA004-B
Supplier:  TEKNOMEK