Human TGF beta 1 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection an tibody, Streptavidin-AP, recombinant human LAP ELISA standard, and standard reconstitution buffer. This matched antib 1 * 1.920 Tests
Catalog # ANTIA270419-2096
Supplier: Antibodies.com