Rat IL-22 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IL-22 ELISA standard, and standard reconstitution buffer. This matched antibody p 1 * 1.920 Tests
Catalog # ANTIA270455-2096
Supplier: ANTIBODIES.COM