Order Entry
Slovenia
ContactUsLinkComponent
Rat Complement C3d ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Complement C3d in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326439-96
Supplier:  ANTIBODIES.COM
New Product
Rat Complement C3d ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Complement C3d in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samp 1 * 96 Tests
Catalog # ANTIA326439-96
Supplier:  ANTIBODIES.COM