Order Entry
Slovenia
ContactUsLinkComponent
This Aspartate Aminotransferase Activity Assay Kit is a quantitative assay kit for measuring Aspartate Aminotransferase activity in serum, plasma, animal and plant tissues, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319663-96
Supplier:  Antibodies.com
New Product
This Aspartate Aminotransferase Activity Assay Kit is a quantitative assay kit for measuring Aspartate Aminotransferase activity in serum, plasma, animal and plant tissues, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319663-96
Supplier:  Antibodies.com