Order Entry
Slovenia
ContactUsLinkComponent
This Phosphoenolpyruvate Carboxykinase / PEPCK Activity Assay Kit is a quantitative assay kit for measuring Phosphoenolpyruvate Carb oxykinase / PEPCK activity in animal tissue, plant tissue, cells, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319715-48
Supplier:  Antibodies.com
New Product
This Phosphoenolpyruvate Carboxykinase / PEPCK Activity Assay Kit is a quantitative assay kit for measuring Phosphoenolpyruvate Carb oxykinase / PEPCK activity in animal tissue, plant tissue, cells, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319715-48
Supplier:  Antibodies.com