Order Entry
Slovenia
ContactUsLinkComponent
This Glucose-6-Phosphate Dehydrogenase Activity Assay Kit is a quantitative assay kit for measuring Glucose-6-Phosphate Dehydrogenas e activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319681-48
Supplier:  Antibodies.com
New Product
This Glucose-6-Phosphate Dehydrogenase Activity Assay Kit is a quantitative assay kit for measuring Glucose-6-Phosphate Dehydrogenas e activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 48 Tests
Catalog # ANTIA319681-48
Supplier:  Antibodies.com