Order Entry
Slovenia
ContactUsLinkComponent
Human C6 ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determination of human C6 in se rum and plasma. 1 * 96 Tests
Catalog # ANTIA324314-96
Supplier:  Antibodies.com
New Product
Human C6 ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determination of human C6 in se rum and plasma. 1 * 96 Tests
Catalog # ANTIA324314-96
Supplier:  Antibodies.com