Order Entry
Slovenia
ContactUsLinkComponent
This Tissue and Blood Alkaline Phosphatase Activity Assay Kit is a quantitative assay kit for measuring Alkaline Phosphatase activit y in serum, plasma, animal tissues, and urine. 1 * 96 Tests
  ANTIA319746-96
New Product
This Tissue and Blood Alkaline Phosphatase Activity Assay Kit is a quantitative assay kit for measuring Alkaline Phosphatase activit y in serum, plasma, animal tissues, and urine. 1 * 96 Tests
  ANTIA319746-96