Order Entry
Slovenia
ContactUsLinkComponent
This Oxygen Radical Antioxidant Capacity Fluorometric Assay Kit is a quantitative assay kit for measuring Oxygen Radical Antioxidant Capacity in animal and plant tissues, cells, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319769-48
Supplier:  Antibodies.com
New Product
This Oxygen Radical Antioxidant Capacity Fluorometric Assay Kit is a quantitative assay kit for measuring Oxygen Radical Antioxidant Capacity in animal and plant tissues, cells, bacteria, serum, and plasma. 1 * 48 Tests
Catalog # ANTIA319769-48
Supplier:  Antibodies.com