Order Entry
Slovenia
ContactUsLinkComponent
This Thioredoxin Peroxidase / Periredoxin 4 Assay Kit is a quantitative assay kit for measuring Thioredoxin Peroxidase / Peroxiredox in 4 activity in serum, plasma, animal tissues, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319744-96
Supplier:  Antibodies.com
New Product
This Thioredoxin Peroxidase / Periredoxin 4 Assay Kit is a quantitative assay kit for measuring Thioredoxin Peroxidase / Peroxiredox in 4 activity in serum, plasma, animal tissues, cells, and bacteria. 1 * 96 Tests
Catalog # ANTIA319744-96
Supplier:  Antibodies.com