Order Entry
Slovenia
ContactUsLinkComponent
This Na+/K+-ATPase Activity Assay Kit is a quantitative assay kit for measuring Na+/K+-A TPase activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319707-96
Supplier:  Antibodies.com
New Product
This Na+/K+-ATPase Activity Assay Kit is a quantitative assay kit for measuring Na+/K+-A TPase activity in serum, plasma, animal and plant tissues, cells, cell supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319707-96
Supplier:  Antibodies.com