This Malate Dehydrogenase (NADP+/NADPH) Activity Assay Kit is a quantitative assay kit for measuring Malate Dehydrogenase (NADP+/NAD PH) activity in animal and plant tissues, cells, bacteria, serum, plasma, and other samples 1 * 48 Tests
Catalog # ANTIA319711-48
Supplier: Antibodies.com