Order Entry
Slovenia
ContactUsLinkComponent
This gamma Glutamyl Transpeptidase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Transpeptidase activi ty in serum, plasma, animal and plant tissues, cells, and bacteria. 1 * 96 Tests
  ANTIA319678-96
New Product
This gamma Glutamyl Transpeptidase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Transpeptidase activi ty in serum, plasma, animal and plant tissues, cells, and bacteria. 1 * 96 Tests
  ANTIA319678-96