Order Entry
Slovenia
ContactUsLinkComponent
This gamma Glutamyl Cysteine Ligase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Cysteine Ligase acti vity in serum, plasma, animal and plant tissues, cells, bacteria, and fungi. 1 * 96 Tests
Catalog # ANTIA319760-96
Supplier:  Antibodies.com
New Product
This gamma Glutamyl Cysteine Ligase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Cysteine Ligase acti vity in serum, plasma, animal and plant tissues, cells, bacteria, and fungi. 1 * 96 Tests
Catalog # ANTIA319760-96
Supplier:  Antibodies.com