Mouse Prostate Secretory Protein/PSP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Prostate Secretory Protein/PSP in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7324-48
Supplier: Antibodies.com