Order Entry
Slovenia
ContactUsLinkComponent
Mouse Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Myosin Light Chain Kinase/MLCK in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
  ANTIA74227-96
New Product
Mouse Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of mouse Myosin Light Chain Kinase/MLCK in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
  ANTIA74227-96