Order Entry
Slovenia
ContactUsLinkComponent
Human Thyroid Hormone Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Thyroid Hormone Receptor alpha in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6507-48
Supplier:  Antibodies.com
New Product
Human Thyroid Hormone Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Thyroid Hormone Receptor alpha in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6507-48
Supplier:  Antibodies.com