Order Entry
Slovenia
ContactUsLinkComponent
Daisogel SP-100-10-C4-Phenyl-HP, 10 µm, 300 x 4.6 mm 1 * 1 items
Catalog # MAISDA10.4PH.S3046
Supplier:  DR MAISCH
New Product
Daisogel SP-100-10-C4-Phenyl-HP, 10 µm, 300 x 4.6 mm 1 * 1 items
Catalog # MAISDA10.4PH.S3046
Supplier:  DR MAISCH