Order Entry
Slovenia
ContactUsLinkComponent
Set electronic control unit E Plus Gen.2 For MIKRO 185, EBA 200, C, S, HAEMATOKRIT 200, BOECO MC-24, SC-8, HC-240 Consisting of: 1x electronic control unit E Plus (Gen.2) 1 * 1 items
Catalog # HETTE4133-S
Supplier:  Hettich
New Product
Set electronic control unit E Plus Gen.2 For MIKRO 185, EBA 200, C, S, HAEMATOKRIT 200, BOECO MC-24, SC-8, HC-240 Consisting of: 1x electronic control unit E Plus (Gen.2) 1 * 1 items
Catalog # HETTE4133-S
Supplier:  Hettich