Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Furonitrile 98%

2-Furonitrile 98%

Bottle
Catalog # APOSOR30131-100G
New Product
Supplier:   Apollo Scientific
2-Furonitrile 98%
Catalog # APOSOR30131-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  617-90-3
2-Furonitrile 98%

Bottle

CAS Number:  617-90-3
Price
310,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Cyanofuran
  • CAS Number:617-90-3
  • Formula:C₅H₃NO
  • MDL Number:MFCD00003223
  • Molecular Weight:93.08 g/mol
  • Boiling Pt:146…148 °C
  • Density:1.064 g/cm³ (25 °C)
  • Flash Pt:35 °C
  •  :Density: 1.085 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.