POMPE DC B 24V N86KNDC B 1 * 1 items
Catalog # KNFAN86KNDCB
Supplier: KNF
New Product
Frequently Bought Together
VWR® Reagent Reservoirs
Supplier: VWR Collection
Accessories for Diaphragm Pumps
Supplier: KNF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Accessories for Hei-VAP series
Supplier: Heidolph Instruments GmbH & Co.KG
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK