Order Entry
Slovenia
ContactUsLinkComponent
Comfort rack for chest freezers for 14 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 774x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24520B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 14 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 774x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24520B
Supplier:  TENAK