Order Entry
Slovenia
ContactUsLinkComponent
Classic rack for chest freezers for 15 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 634x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21387B10
Supplier:  TENAK
New Product
Classic rack for chest freezers for 15 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 634x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21387B10
Supplier:  TENAK