Order Entry
Slovenia
ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 9x4=36 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 325x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE11138B10
 :  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 9x4=36 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 325x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE11138B10
 :  TENAK