Order Entry
Slovenia
ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 6x4=24 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 217x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE11132B10
 :  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 6x4=24 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 217x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE11132B10
 :  TENAK