Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Justiergas 6D 20,9 % O2 / N2 103 Liter Stahlzylinder, nicht wiederbefüllbar 1 * 1 ST
Catalog # CALGC003484
Supplier:
CALGAZ
New Product
Expand image
Justiergas 6D 20,9 % O2 / N2 103 Liter Stahlzylinder, nicht wiederbefüllbar 1 * 1 ST
Catalog # CALGC003484
Supplier:
CALGAZ
New Product
Frequently Bought Together
VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions
Supplier: Hanna
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
FREEZING POINT STD CRYOSCOPE 621 100ML 1 * 1 items
Supplier: REAGECON
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
pH buffer, single-use, SINGLET™
Supplier: Hach