Order Entry
Slovenia
ContactUsLinkComponent
MICROBIOLOGICAL SAFETY CABINET CLASS II FOR CYTOTOXIC, SAFEMATE CYTO 1.2, DIN12980 COMPLIANT 1 * 1 items
Catalog # EUCLLY84000
Supplier:  BioAir S.p.A.
New Product
MICROBIOLOGICAL SAFETY CABINET CLASS II FOR CYTOTOXIC, SAFEMATE CYTO 1.2, DIN12980 COMPLIANT 1 * 1 items
Catalog # EUCLLY84000
Supplier:  BioAir S.p.A.