Order Entry
Slovenia
ContactUsLinkComponent
This Uric Acid Assay Kit is a quantitative assay kit for measuring Uric Acid in serum, plasma, urine, and animal tissues. 1 * 96 Tests
Catalog # ANTIA319754-96
Supplier:  Antibodies.com
New Product
This Uric Acid Assay Kit is a quantitative assay kit for measuring Uric Acid in serum, plasma, urine, and animal tissues. 1 * 96 Tests
Catalog # ANTIA319754-96
Supplier:  Antibodies.com