Order Entry
Slovenia
ContactUsLinkComponent
This Total Cholesterol Assay Kit is a quantitative assay kit for measuring Total Cholesterol in serum, plasma, animal tissues, cells , and bacteria. 1 * 48 Tests
Catalog # ANTIA319750-48
Supplier:  Antibodies.com
New Product
This Total Cholesterol Assay Kit is a quantitative assay kit for measuring Total Cholesterol in serum, plasma, animal tissues, cells , and bacteria. 1 * 48 Tests
Catalog # ANTIA319750-48
Supplier:  Antibodies.com