Order Entry
Slovenia
ContactUsLinkComponent
This Superoxide Dismutases Activity Assay Kit is a quantitative assay kit for measuring Superoxide Dismutase activity in serum, plas ma, animal and plant tissues, and cells. 1 * 96 Tests
Catalog # ANTIA319741-96
Supplier:  Antibodies.com
New Product
This Superoxide Dismutases Activity Assay Kit is a quantitative assay kit for measuring Superoxide Dismutase activity in serum, plas ma, animal and plant tissues, and cells. 1 * 96 Tests
Catalog # ANTIA319741-96
Supplier:  Antibodies.com