Order Entry
Slovenia
ContactUsLinkComponent
This Pyruvate Acid Assay Kit is a quantitative assay kit for measuring Pyruvic Acid in animal and plant tissues, cells, bacteria, pl asma, and serum. 1 * 96 Tests
Catalog # ANTIA319724-96
Supplier:  Antibodies.com
New Product
This Pyruvate Acid Assay Kit is a quantitative assay kit for measuring Pyruvic Acid in animal and plant tissues, cells, bacteria, pl asma, and serum. 1 * 96 Tests
Catalog # ANTIA319724-96
Supplier:  Antibodies.com