This Lactate Assay Kit is a quantitative assay kit for measuring Lactate in animal and plant tissues, cells, plasma, serum, and othe r liquid samples. 1 * 96 Tests
Catalog # ANTIA319694-96
Supplier: Antibodies.com