Order Entry
Slovenia
ContactUsLinkComponent
This Hydrogen Peroxide Assay Kit is a quantitative assay kit for measuring Hydrogen Peroxide in serum, plasma, urine, animal and pla nt tissues, cells, and bacteria. 1 * 480 Tests
Catalog # ANTIA319692-480
Supplier:  Antibodies.com
New Product
This Hydrogen Peroxide Assay Kit is a quantitative assay kit for measuring Hydrogen Peroxide in serum, plasma, urine, animal and pla nt tissues, cells, and bacteria. 1 * 480 Tests
Catalog # ANTIA319692-480
Supplier:  Antibodies.com