Order Entry
Slovenia
ContactUsLinkComponent
This Glucose Assay Kit is a quantitative assay kit for measuring Glucose in animal and plant tissues, cells, bacteria, serum, plasma , urine, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA319679-96
Supplier:  Antibodies.com
New Product
This Glucose Assay Kit is a quantitative assay kit for measuring Glucose in animal and plant tissues, cells, bacteria, serum, plasma , urine, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA319679-96
Supplier:  Antibodies.com