Order Entry
Slovenia
ContactUsLinkComponent
This Creatine Kinase Activity Assay Kit is a quantitative assay kit for measuring Creatine Kinase activity in animal tissues, cells, and serum. 1 * 96 Tests
Catalog # ANTIA319671-96
Supplier:  Antibodies.com
New Product
This Creatine Kinase Activity Assay Kit is a quantitative assay kit for measuring Creatine Kinase activity in animal tissues, cells, and serum. 1 * 96 Tests
Catalog # ANTIA319671-96
Supplier:  Antibodies.com