VIOLET RED BILE AGAR MUG 1 * 100 g
Catalog # LIOF620187
Supplier: LIOFILCHEM
New Product
Frequently Bought Together
Culture media in bottles
Supplier: LIOFILCHEM
POTATO DEXTROSE BROTH 1 * 100 g
Supplier: LIOFILCHEM
VWR® Culture Media, Ready To Use, In Tubes
Supplier: VWR Chemicals
Antimicrobial susceptibility disks
Supplier: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Haemophilus influenzae</i> NCTC™ 8468™
Supplier: THERMO FISHER DIAGNOSTICS
TEST IDENTIFICAZ./CLASSIFICA- ZIONE 1 * 1 items
Supplier: LIOFILCHEM
CultiControl freeze-dried microorganism strains derived from NCTC® cultures
Supplier: LIOFILCHEM
PLATE COUNT AGAR 1 * 100 g
Supplier: LIOFILCHEM
VWR®, Pre-weighed dehydrated culture media in sachets
Supplier: VWR Chemicals
HAEMOGLOBIN SOLUBLE POWDER 1 * 400 g
Supplier: THERMO FISHER DIAGNOSTICS