Order Entry
Slovenia
ContactUsLinkComponent
Agilent HPLC with MWR Detector - 1260 Infinity II includes: 1260 Infinity II Quaternary Pump VL, HPLC System Tool Kit 1260 Infinity II, Active Inlet Valve (AIV),Poroshell 120 EC-C18, 4.6x100mm, 2.7um, 1260 Infinity II Vialsampler, Standard drawer (6 1 * 1 SET
Catalog # IBSAHPLC1260INFMWD
Supplier:  IBS Stock Items
New Product
Agilent HPLC with MWR Detector - 1260 Infinity II includes: 1260 Infinity II Quaternary Pump VL, HPLC System Tool Kit 1260 Infinity II, Active Inlet Valve (AIV),Poroshell 120 EC-C18, 4.6x100mm, 2.7um, 1260 Infinity II Vialsampler, Standard drawer (6 1 * 1 SET
Catalog # IBSAHPLC1260INFMWD
Supplier:  IBS Stock Items