Order Entry
Slovenia
ContactUsLinkComponent
Agilent HPLC with MWD + FLD Detector - 1260 Infinity II includes: 1260 Infinity II Quaternary Pump VL, HPLC System Tool Kit 1260 Infinity II , Active Inlet Valve (AIV), Poroshell 120 EC-C18, 4.6x100mm, 2.7um, 1260 Infinity II Vialsampler, Standard d 1 * 1 SET
Catalog # IBSAHPLC1260INMWDF
Supplier:  IBS Stock Items
New Product
Agilent HPLC with MWD + FLD Detector - 1260 Infinity II includes: 1260 Infinity II Quaternary Pump VL, HPLC System Tool Kit 1260 Infinity II , Active Inlet Valve (AIV), Poroshell 120 EC-C18, 4.6x100mm, 2.7um, 1260 Infinity II Vialsampler, Standard d 1 * 1 SET
Catalog # IBSAHPLC1260INMWDF
Supplier:  IBS Stock Items