Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Mouse Recombinant TECK (CCL25)

Mouse Recombinant TECK (CCL25)

25 µG
Catalog # STMC78180.1
Mouse Recombinant TECK (CCL25)
Catalog # STMC78180.1
CAS Number:  
Mouse Recombinant TECK (CCL25)
Mouse Recombinant TECK (CCL25)

25 µG

CAS Number:  
Price
318,00 €
Log in for availability
1 * 25 µG
Product Details & Documents

Thymus-expressed chemokine (TECK), or CCL25, is a member of the CC family of chemokines that regulates the movement of lymphocytes in the thymus and in the small intestine. TECK induces chemoattraction by binding the chemokine receptor CCR9, which is expressed on immature pre-T cells and thymocytes (Youn et al.; Uehara et al.). In the thymus, TECK is produced by stromal cells, whereas in the small intestine TECK is primarily produced by epithelial cells (Vicari et al.; Bowman et al.; Kunkel et al.). CCR9 is a G protein-coupled receptor and is expressed on most thymocytes, but not on natural killer cells, monocytes, eosinophils, basophils, and neutrophils (Wu et al). In Jurkat cells, binding of TECK to CCL9 has been shown to increase levels of intracellular calcium (Cheng-Rong et al). TECK/CCR9 signaling has also been linked to many cancers, as these molecules have been shown to mediate anti-apoptotic processes by activating the PI3K/AKT signaling pathway, weakening the effect of cytotoxic T cells by regulating STAT signaling. Additionally, TECK induces metastasis by increasing the expression of MMP2 and MMP9 (Tu et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    25 µG
  • Cat. No.:
    STMC78180.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TECK (CCL25)
  • Purity:
    ≥98%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.