Streptococcus pneumoniae Identification Kit 1 * 1 KIT
Catalog # MSPPR30033
Supplier: MARKET SOURCE PART PROCESS
Streptococcus pneumoniae Identification Kit 1 * 1 KIT
Catalog # MSPPR30033
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
NANOCOLOR® Photometric Tube Tests Kits for Ammonium
Supplier: MACHEREY-NAGEL
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, quaternary ammonium compounds (QUAT), QUANTOFIX®
Supplier: MACHEREY-NAGEL
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS