RET Split Probe is designed for the qualitative detection of translocations involving the human RET gene at 10q11.21 in formalin-fix ed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH). 1 * 100 µl
Catalog # ABNOCS0018.100UL
Supplier: Abnova
Frequently Bought Together