PEFORM DISINFECTANT 40g BT 1 * 60 ST
Catalog # SCUE70001858
Supplier: SCHUELKE MAYR
Frequently Bought Together
Interfold and roll hand towels, Tork
Supplier: Essity
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
VWR®, Nitrile Gloves
Supplier: VWR Collection
Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE
Silicone keyboard for the medical sector, GCQ CleanType® Easy Protect
Supplier: GETT GERATETECHNIK