Order Entry
Slovenia
ContactUsLinkComponent
CX23RTFS2-1-2 upright microscope set for transmitted light observation. CX23 microscope frame with integrated LED illumination, quadruple nosepiece, fixed mechanical stage (174 x 89 mm, travelling range 76 x 30 mm) with right hand low drive control a 1 * 1 ST
Catalog # OLYMN5731300
Supplier:  OLYMPUS OPTICAL
CX23RTFS2-1-2 upright microscope set for transmitted light observation. CX23 microscope frame with integrated LED illumination, quadruple nosepiece, fixed mechanical stage (174 x 89 mm, travelling range 76 x 30 mm) with right hand low drive control a 1 * 1 ST
Catalog # OLYMN5731300
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together