0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
DENSITY STANDARD 1,631 G/ML AT 15°C 1 * 1.000 mL
ACSDDNS16300.L1
:
CPACHEM
DENSITY STANDARD 1,631 G/ML AT 15°C 1 * 1.000 mL
ACSDDNS16300.L1
:
CPACHEM
Frequently Bought Together
PARACETAMOL IMPURITY J 1 * 15 mg
: EDQM
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution USP Test Solution (TS) 34
: VWR Chemicals
Imidaclothiz 1 * 25 mg
: EHRENSTORFER
1-Bromo-2-chloroethane
: EHRENSTORFER
Ursolic Acid, Reference Standard
: USP
Buffer Solutions
: Thermo Fisher Scientific
(±)-1-Butoxy-2-propanol
: EHRENSTORFER
Lactofen certified 1 * 0,1 g
: EHRENSTORFER