FELODIPINE FOR PEAK IDENTIFICATION 1 * 15 mg
Catalog # EDQMY0002366
Supplier: EDQM
Frequently Bought Together
VWR® ARISTAR®, Multi-Element Calibration Standard 2 in dil. Nitric Acid for ICP-MS
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets
Supplier: Mettler - Toledo
Ammonia solution 17% Reag. Ph. Eur. 1004701
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals